3-Iodo-1-phenylpropane 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A612779-1G A612779-250MG A612779-25G A612779-5G
77242-702EA 27 USD
77242-702 77242-704 77242-706 77242-708
3-Iodo-1-phenylpropane 98% stabilized
3-Iodo-1-phenylpropane

Precaución: for Research Use Only

Formula: C₉H₁₁I
MDL Number: MFCD00019036
CAS Number: 4119-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR