(S)-[1,1'-Binaphthalene]-2,2'-diylbis[1,1-diphenyl-1,1'-phosphineoxide] ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A612494-100MG
A612494-1G
A612494-250MG
A612494-5G
77254-326EA
20.6
USD
77254-326
77254-328
77254-330
77254-332
(S)-[1,1'-Binaphthalene]-2,2'-diylbis[1,1-diphenyl-1,1'-phosphineoxide] ≥98%, ee 99%
(S)-[1,1'-Binaphthalene]-2,2'-diylbis[1,1-diphenyl-1,1'-phosphineoxide]
Precaución: for Research Use Only
Formula:
C₄₄H₃₂O₂P₂ MW: 654.67 g/mol |
MDL Number:
MFCD00822644 CAS Number: 94041-18-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...