2-Mesitylenesulfonyl chloride 95%
Supplier: Ambeed
|
Danger
|
A612400-100G
A612400-10G
A612400-25G
A612400-5G
77254-318EA
47
USD
77254-318
77254-320
77254-322
77254-324
2-Mesitylenesulfonyl chloride 95%
2-Mesitylenesulphonyl chloride
Precaución: for Research Use Only
Formula:
C₉H₁₁ClO₂S |
MDL Number:
MFCD00007434 CAS Number: 773-64-8 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...