3-(Pentafluorophenyl)propionic acid 95%

Supplier: Ambeed
Danger

Synonyms: 3-(Perfluorophenyl)propanoic acid

A612292-1G A612292-250MG A612292-5G
77280-318EA 134.93 USD
77280-318 77280-320 77280-322
3-(Pentafluorophenyl)propionic acid 95%
3-(Pentafluorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₅F₅O₂
MW: 240.13 g/mol
MDL Number: MFCD00020516
CAS Number: 2002-92-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR