4-Methylumbelliferyl sulfate potassium salt 98%

Supplier: Ambeed
Warning

Synonyms: Potassium 4-methyl-2-oxo-2H-chromen-7-yl sulphate, 4-Methylumbelliferyl sulphate potassium

A612240-100MG A612240-250MG A612240-5G A612240-1G
77292-852EA 14 USD
77292-852 77617-558 77617-560 77749-260
4-Methylumbelliferyl sulfate potassium salt 98%
4-Methylumbelliferyl sulfate potassium salt

Precaución: for Research Use Only

Formula: C₁₀H₇KO₆S
MW: 294.318 g/mol
MDL Number: MFCD00016970
CAS Number: 15220-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR