9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole 98%

Supplier: Ambeed
Warning

Synonyms: 9-(3-(Dibenzo[b,d]furan-2-yl)phenyl)-9H-carbazole

A612079-100MG A612079-250MG
77242-698EA 36.05 USD
77242-698 77242-700
9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole 98%
9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole

Precaución: for Research Use Only

Formula: C₃₀H₁₉NO
MW: 409.49 g/mol
MDL Number: MFCD30478530
CAS Number: 1338446-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR