8,10-Dihydro-s-indaceno[1,2-b:6,7-b']dithiophene 97%

Supplier: Ambeed
Warning

A611907-100MG A611907-1G A611907-250MG
77280-312EA 15 USD
77280-312 77280-314 77280-316
8,10-Dihydro-s-indaceno[1,2-b:6,7-b']dithiophene 97%
8,10-Dihydro-s-indaceno[1,2-b:6,7-b']dithiophene

Precaución: for Research Use Only

Formula: C₁₆H₁₀S₂
MDL Number: MFCD31560770
CAS Number: 1209012-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR