2-(4-Bromo-1H-indol-3-yl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromoindole-3-acetic Acid

A611228-1G A611228-250MG A611228-5G
77242-692EA 110.21 USD
77242-692 77242-694 77242-696
2-(4-Bromo-1H-indol-3-yl)acetic acid 98%
2-(4-Bromo-1H-indol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
MW: 254.0800 g/mol
MDL Number: MFCD09751717
CAS Number: 89245-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR