2'-Chloro-4'-fluoroacetanilide 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-Chloro-4-fluorophenyl)acetamide

A610917-25G A610917-5G
77266-810EA 72.1 USD
77266-810 77266-812
2'-Chloro-4'-fluoroacetanilide 98%
2'-Chloro-4'-fluoroacetanilide

Precaución: for Research Use Only

Formula: C₈H₇ClFNO
MW: 187.6 g/mol
MDL Number: MFCD00042594
CAS Number: 399-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR