4-Bromo-N-methylaniline 98%

Supplier: Ambeed
Warning

A610421-10G A610421-1G A610421-250MG A610421-5G
77292-844EA 71.07 USD
77292-844 77292-846 77292-848 77292-850
4-Bromo-N-methylaniline 98%
4-Bromo-N-methylaniline

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD03030391
CAS Number: 6911-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR