4,5-Dichloroisatin 95%

Supplier: Ambeed
Warning

A610209-100MG A610209-1G A610209-250MG
77292-838EA 33.99 USD
77292-838 77292-840 77292-842
4,5-Dichloroisatin 95%
4,5-Dichloroisatin

Precaución: for Research Use Only

Formula: C₈H₃Cl₂NO₂
MW: 216.02 g/mol
MDL Number: MFCD00185014
CAS Number: 1677-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR