(R)-Ethyl 6-(4-phenyl-4,5-dihydrooxazol-2-yl)nicotinate 98%
Supplier: Ambeed
|
Warning
|
A608830-100MG
A608830-1G
A608830-250MG
77266-804EA
150
USD
77266-804
77266-806
77266-808
(R)-Ethyl 6-(4-phenyl-4,5-dihydrooxazol-2-yl)nicotinate 98%
(R)-Ethyl 6-(4-phenyl-4,5-dihydrooxazol-2-yl)nicotinate
Precaución: for Research Use Only
Formula:
C₁₇H₁₆N₂O₃ |
CAS Number:
1085431-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...