4-(2-Nitrophenyl)thiazole-2-amine 97%

Supplier: Ambeed
Warning

A608824-100MG A608824-1G A608824-250MG
77292-826EA 44.29 USD
77292-826 77292-828 77292-830
4-(2-Nitrophenyl)thiazole-2-amine 97%
4-(2-Nitrophenyl)thiazole-2-amine

Precaución: for Research Use Only

Formula: C₉H₇N₃O₂S
MW: 221.24 g/mol
MDL Number: MFCD02352318
CAS Number: 90323-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR