MRT67307 98%

Supplier: Ambeed
Warning

Synonyms: N-[3-[[5-cyclopropyl-2-[3-(morpholin-4-ylmethyl)anilino]pyrimidin-4-yl]amino]propyl]cyclobutanecarboxamide, N-(3-((5-Cyclopropyl-2-((3-(morpholinomethyl)phenyl)amino)pyrimidin-4-yl)amino)propyl)cyclobutanecarboxamide

A608717-10MG A608717-1MG A608717-5MG
77242-684EA 63 USD
77242-684 77242-686 77242-688
MRT67307 98%
MRT67307

Precaución: for Research Use Only

Formula: C₂₆H₃₆N₆O₂
MW: 464.62 g/mol
MDL Number: MFCD19443643
CAS Number: 1190378-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR