4-Chloro-N-(6-chloropyridin-3-yl)benzamide 99%

Supplier: Ambeed
Warning

A608671-10MG A608671-1MG A608671-25MG A608671-5MG
77242-676EA 35.02 USD
77242-676 77242-678 77242-680 77242-682
4-Chloro-N-(6-chloropyridin-3-yl)benzamide 99%
4-Chloro-N-(6-chloropyridin-3-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂N₂O
MW: 267.11 g/mol
MDL Number: MFCD05669575
CAS Number: 325457-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR