6-Bromo-7-hydroxy-2,2-dimethyl-4H-benzo[d][1,3]dioxin-4-one 97%
Supplier: Ambeed
|
Warning
|
A608661-250MG
A608661-1G
A608661-100MG
77254-316EA
84.46
USD
77254-316
77747-253
77747-252
6-Bromo-7-hydroxy-2,2-dimethyl-4H-benzo[d][1,3]dioxin-4-one 97%
6-Bromo-7-hydroxy-2,2-dimethyl-4H-benzo[d][1,3]dioxin-4-one
Precaución: for Research Use Only
Formula:
C₁₀H₉BrO₄ MW: 273.08 g/mol |
CAS Number:
531501-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...