(±)-1-Phenoxy-2-propanol 99%

Supplier: Ambeed
Warning

Synonyms: 1-Phenoxy-2-propanol, Propylene Phenoxytol, (±)-Monopropylene glycol phenyl ether

A608603-1000G A608603-100G A608603-25G A608603-500G
77242-668EA 26.78 USD
77242-668 77242-670 77242-672 77242-674
(±)-1-Phenoxy-2-propanol 99%
(±)-1-Phenoxy-2-propanol

Precaución: for Research Use Only

Formula: C₆H₅OCH₂CH(OH)CH₃
MW: 152.19 g/mol
MDL Number: MFCD00016861
CAS Number: 770-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR