2,3,5-Trimethylaniline 97%

Supplier: Ambeed
Warning

A608417-100MG A608417-1G A608417-250MG
77266-796EA 35.02 USD
77266-796 77266-798 77266-800
2,3,5-Trimethylaniline 97%
2,3,5-Trimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.20 g/mol
MDL Number: MFCD01863780
CAS Number: 767-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR