Boc-Thz-OH 98%

Supplier: Ambeed
Warning

Synonyms: Boc-L-thiazolidine-4-carboxylic acid

A608127-100G A608127-10G A608127-1G A608127-25G A608127-500G A608127-5G
77254-304EA 83 USD
77254-304 77254-306 77254-308 77254-310 77254-312 77254-314
Boc-Thz-OH 98%
Boc-Thz-OH

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄S
MDL Number: MFCD00077002
CAS Number: 51077-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR