(1R,1'R)-1,1'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2'-bis[(R)-(dimethylamino)phenylmethyl]ferrocene 98%

Supplier: Ambeed
Warning

A606870-100MG A606870-250MG A606870-1G
77254-300EA 73 USD
77254-300 77254-302 77747-251
(1R,1'R)-1,1'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2'-bis[(R)-(dimethylamino)phenylmethyl]ferrocene 98%
(1R,1'R)-1,1'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2,2'-bis[(R)-(dimethylamino)phenylmethyl]ferrocene

Precaución: for Research Use Only

Formula: C₆₄H₇₅FeN₂O₄P₂
MDL Number: MFCD04117704
CAS Number: 876608-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR