6-Amino-5-chloro-2-methylnicotinonitrile 97%

Supplier: Ambeed
Danger

A606814-100MG A606814-1G A606814-250MG A606814-50MG
77242-660EA 48.41 USD
77242-660 77242-662 77242-664 77242-666
6-Amino-5-chloro-2-methylnicotinonitrile 97%
6-Amino-5-chloro-2-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.59 g/mol
MDL Number: MFCD26129201
CAS Number: 1504581-92-3
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR