2-Bromo-1-(bromomethyl)-3-methylbenzene 97%

Supplier: Ambeed
Danger

A606242-250MG A606242-5G A606242-1G
77266-776EA 48.41 USD
77266-776 77572-966 77749-244
2-Bromo-1-(bromomethyl)-3-methylbenzene 97%
2-Bromo-1-(bromomethyl)-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂
MW: 263.95 g/mol
MDL Number: MFCD18392013
CAS Number: 66790-58-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR