3-Hydroxy-1-(4-methoxyphenyl)quinazoline-2,4(1H,3H)-dione 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
FEN1-IN-3
A605561-10MG
A605561-1MG
A605561-25MG
A605561-50MG
A605561-5MG
77266-766EA
71.07
USD
77266-766
77266-768
77266-770
77266-772
77266-774
3-Hydroxy-1-(4-methoxyphenyl)quinazoline-2,4(1H,3H)-dione 98%
3-Hydroxy-1-(4-methoxyphenyl)quinazoline-2,4(1H,3H)-dione
Precaución: for Research Use Only
Formula:
C₁₅H₁₂N₂O₄ MW: 284.26 g/mol |
CAS Number:
2109805-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...