N-(5-(6-Chloro-2,2-difluorobenzo[d][1,3]dioxol-5-yl)pyrazin-2-yl)-2-fluoro-6-methylbenzamide 97%

Supplier: Ambeed
Warning

A605542-10MG A605542-1MG A605542-50MG A605542-5MG
77254-292EA 123 USD
77254-292 77254-294 77254-296 77254-298
N-(5-(6-Chloro-2,2-difluorobenzo[d][1,3]dioxol-5-yl)pyrazin-2-yl)-2-fluoro-6-methylbenzamide 97%
N-(5-(6-Chloro-2,2-difluorobenzo[d][1,3]dioxol-5-yl)pyrazin-2-yl)-2-fluoro-6-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₉H₁₁ClF₃N₃O₃
MDL Number: MFCD30489478
CAS Number: 1713240-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR