2-Acetyl-5-bromothiophene 98%

Supplier: Ambeed
Warning

A605521-10G A605521-25G A605521-5G A605521-100G
77292-816EA 10 USD
77292-816 77292-818 77292-820 77749-243
2-Acetyl-5-bromothiophene 98%
2-Acetyl-5-bromothiophene

Precaución: for Research Use Only

Formula: C₆H₅BrOS
MDL Number: MFCD00014528
CAS Number: 5370-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR