4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4'-[(6-Acryloyloxy)hexyloxy]-4-biphenylcarbonitrile, 6-[(4'-Cyanobiphenyl-4-yl)oxy]hexyl acrylate, Acrylic acid 6-[(4'-Cyano-[1,1'-biphenyl]-4-yl)oxy]hexyl ester, 6-[(4'-Cyano-[1,1'-biphenyl]-4-yl)oxy]hexyl acrylate, 6-((4'-Cyano-[1,1'-biphenyl]-4-yl)oxy)hexyl acrylate

A605379-1G A605379-250MG A605379-25G A605379-5G
77254-284EA 35 USD
77254-284 77254-286 77254-288 77254-290
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl 98%
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl

Precaución: for Research Use Only

Formula: C₂₂H₂₃NO₃
MDL Number: MFCD16293619
CAS Number: 89823-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR