3-Bromo-5-methylthiophene-2-carbaldehyde 97%

Supplier: Ambeed
Warning

A604770-100MG A604770-1G
77280-278EA 33.99 USD
77280-278 77280-280
3-Bromo-5-methylthiophene-2-carbaldehyde 97%
3-Bromo-5-methylthiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₅BrOS
MW: 205.07 g/mol
MDL Number: MFCD01859869
CAS Number: 36155-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR