3-Methylpyridine-2-sulfonamide 95%

Supplier: Ambeed
Warning

A604749-100MG A604749-1G A604749-250MG
77280-272EA 15 USD
77280-272 77280-274 77280-276
3-Methylpyridine-2-sulfonamide 95%
3-Methylpyridine-2-sulfonamide

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
MDL Number: MFCD11046395
CAS Number: 65938-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR