3-(4-Bromophenyl)prop-2-yn-1-ol 97%

Supplier: Ambeed
Warning

A604625-100MG A604625-1G A604625-250MG
77280-266EA 19.57 USD
77280-266 77280-268 77280-270
3-(4-Bromophenyl)prop-2-yn-1-ol 97%
3-(4-Bromophenyl)prop-2-yn-1-ol

Precaución: for Research Use Only

Formula: C₉H₇BrO
MW: 211.055 g/mol
MDL Number: MFCD01250510
CAS Number: 37614-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR