4-((1,4-Dioxo-1,4-dihydronaphthalen-2-yl)amino)benzenesulfonamide 97%

Supplier: Ambeed
Warning

A604604-10MG A604604-25MG A604604-50MG A604604-5MG
77254-270EA 6 USD
77254-270 77254-272 77254-274 77254-276 77254-278
4-((1,4-Dioxo-1,4-dihydronaphthalen-2-yl)amino)benzenesulfonamide 97%
4-((1,4-Dioxo-1,4-dihydronaphthalen-2-yl)amino)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂O₄S
MW: 328.34 g/mol
MDL Number: MFCD30579194
CAS Number: 19992-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR