(S)-1-Benzyl-3-pyrrolidinol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Benzyl-3-hydroxypyrrolidine, (S)-1-Benzylpyrrolidin-3-ol

A604599-100G A604599-10G A604599-1G A604599-5G
77280-260EA 209.09 USD
77280-260 77280-262 77280-264 77617-532
(S)-1-Benzyl-3-pyrrolidinol 97%
(S)-1-Benzyl-3-pyrrolidinol

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
MDL Number: MFCD00075485
CAS Number: 101385-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR