Sodium 1-hexadecanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Sodium hexadecane-1-sulphonate, Hexadecane-1-sulphonic acid sodium salt

A604515-100G A604515-25G
77242-652EA 37.08 USD
77242-652 77242-654
Sodium 1-hexadecanesulfonate 98%
Sodium 1-hexadecanesulphonate

Precaución: for Research Use Only

Formula: C₁₆H₃₃NaSO₃
MW: 328.49 g/mol
MDL Number: MFCD00007528
CAS Number: 15015-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR