4-(4-Acetoxyphenyl)-2-butanone 96%

Supplier: Ambeed
Warning

Synonyms: Cue-lure, Acetic Acid 4-(3-Oxobutyl)phenyl Ester, 4-(3-Oxobutyl)phenyl Acetate

A604232-25G A604232-5G A604232-100G
77292-812EA 10.3 USD
77292-812 77292-814 77749-241
4-(4-Acetoxyphenyl)-2-butanone 96%
4-(4-Acetoxyphenyl)-2-butanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD00008704
CAS Number: 3572-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR