3-Chlorovanillin 98%

Supplier: Ambeed
Warning

A603809-1G A603809-250MG A603809-25G A603809-5G
77242-632EA 18 USD
77242-632 77242-634 77242-636 77242-638
3-Chlorovanillin 98%
3-Chlorovanillin

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MDL Number: MFCD00016982
CAS Number: 19463-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR