2-Methylquinoline-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Carboxy-2-methyl-1-azanaphthalene, 3-Carboxy-2-methylquinoline
A603696-250MG
A603696-5G
77280-256EA
21.63
USD
77280-256
77280-258
2-Methylquinoline-3-carboxylic acid 97%
2-Methylquinoline-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₉NO₂ MW: 187.195 g/mol |
MDL Number:
MFCD00510998 CAS Number: 635-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...