α,α,α-Trifluoroacetophenone 98%

Supplier: Ambeed
Danger

A603592-25G A603592-5G
77254-266EA 32 USD
77254-266 77254-268
α,α,α-Trifluoroacetophenone 98%
α,α,α-Trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅F₃O
MDL Number: MFCD00000420
CAS Number: 434-45-7
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR