4-(1H-Pyrazol-1-yl)benzenesulfonyl chloride 96%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-[4-(Chlorosulphonyl)phenyl]-1H-pyrazole, 4-(1H-Pyrazol-1-yl)benzene-1-sulphonyl chloride
A603583-1G
A603583-5G
77292-808EA
49.44
USD
77292-808
77292-810
4-(1H-Pyrazol-1-yl)benzenesulfonyl chloride 96%
4-(1H-Pyrazol-1-yl)benzenesulfonyl chloride
Precaución: for Research Use Only
Formula:
C₉H₇ClN₂O₂S MW: 242.682 g/mol |
MDL Number:
MFCD02681937 CAS Number: 18336-39-5 UN: 3261 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...