(S)-2,6-Diamino-N-((5S,8S,11S,14S,17S,20S,23S,26S)-11,20,23-tris((1H-indol-3-yl)methyl)-1,30-diamino-14,17-bis(4-aminobutyl)-8-benzhydryl-5-carbamoyl-7,10,13,16,19,22,25-heptaoxo-6,9,12,15,18,21,24-heptaazatriacontan-26-yl)hexanamide 98%

Supplier: Ambeed
Warning

Synonyms: Ruxotemitide

A603429-10MG A603429-1MG A603429-25MG A603429-2MG A603429-5MG
77266-748EA 180 USD
77266-748 77266-750 77266-752 77266-754 77266-756
(S)-2,6-Diamino-N-((5S,8S,11S,14S,17S,20S,23S,26S)-11,20,23-tris((1H-indol-3-yl)methyl)-1,30-diamino-14,17-bis(4-aminobutyl)-8-benzhydryl-5-carbamoyl-7,10,13,16,19,22,25-heptaoxo-6,9,12,15,18,21,24-heptaazatriacontan-26-yl)hexanamide 98%
(S)-2,6-Diamino-N-((5S,8S,11S,14S,17S,20S,23S,26S)-11,20,23-tris((1H-indol-3-yl)methyl)-1,30-diamino-14,17-bis(4-aminobutyl)-8-benzhydryl-5-carbamoyl-7,10,13,16,19,22,25-heptaoxo-6,9,12,15,18,21,24-heptaazatriacontan-26-yl)hexanamide

Precaución: for Research Use Only

Formula: C₇₈H₁₀₆N₁₈O₉
MDL Number: MFCD30742993
CAS Number: 1345407-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR