6-Butylpyridazin-3-amine 97%

Supplier: Ambeed
Warning

A603282-100MG A603282-1G A603282-250MG A603282-50MG
77266-740EA 94 USD
77266-740 77266-742 77266-744 77266-746
6-Butylpyridazin-3-amine 97%
6-Butylpyridazin-3-amine

Precaución: for Research Use Only

Formula: C₈H₁₃N₃
MDL Number: MFCD30189277
CAS Number: 1699465-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR