4-Amino-2-chlorophenylboronic acid pinacol ester 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-2-chlorophenylboronic acid,pinacolester, 4-Amino-2-chlorophenylboronic acid, piancol ester
A603131-100MG
A603131-1G
A603131-250MG
A603131-5G
77292-798EA
41.2
USD
77292-798
77292-800
77292-802
77292-804
4-Amino-2-chlorophenylboronic acid pinacol ester 96%
4-Amino-2-chlorophenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BClNO₂ MW: 253.54 g/mol |
MDL Number:
MFCD09864184 CAS Number: 877160-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...