2-(3,5-Dimethoxyphenyl)ethanol 95%

Supplier: Ambeed
Warning

A603080-100MG A603080-250MG
77242-622EA 35 USD
77242-622 77242-624
2-(3,5-Dimethoxyphenyl)ethanol 95%
2-(3,5-Dimethoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD09926335
CAS Number: 7417-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR