Dde-Dab(Fmoc)-OH 97%

Supplier: Ambeed
Warning

A602882-100MG A602882-1G A602882-250MG A602882-5G
77242-614EA 15 USD
77242-614 77242-616 77242-618 77242-620
Dde-Dab(Fmoc)-OH 97%
Dde-Dab(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₉H₃₂N₂O₆
MDL Number: MFCD05663743
CAS Number: 1263045-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR