H-D-Phe(4-NO2)-OH.HCl 97%

Supplier: Ambeed
Warning

A602879-1G A602879-250MG A602879-5G
77266-734EA 6.18 USD
77266-734 77266-736 77266-738
H-D-Phe(4-NO2)-OH.HCl 97%
H-D-Phe(4-NO2)-OH.HCl

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂O₄
MW: 246.64 g/mol
MDL Number: MFCD14636441
CAS Number: 147065-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR