1,8-Bis(diphenylphosphino)naphthalene 98%

Supplier: Ambeed
Warning

A602773-1G A602773-250MG
77242-610EA 152 USD
77242-610 77242-612
1,8-Bis(diphenylphosphino)naphthalene 98%
1,8-Bis(diphenylphosphino)naphthalene

Precaución: for Research Use Only

Formula: C₃₄H₂₆P₂
MDL Number: MFCD04117326
CAS Number: 153725-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR