4-Pentylphenyl 4-Methoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybenzoic acid 4-Amylphenyl ester, 4-Amylphenyl 4-Methoxybenzoate, 4-Methoxybenzoic acid 4-Pentylphenyl ester, 4-Pentylphenyl 4-methoxybenzoate

A602721-25G A602721-5G
77292-794EA 56.65 USD
77292-794 77292-796
4-Pentylphenyl 4-Methoxybenzoate 98%
4-Pentylphenyl 4-Methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₉H₂₂O₃
MW: 298.38 g/mol
MDL Number: MFCD08276868
CAS Number: 38444-13-2
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR