2-Methoxybenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A602500-10G A602500-1G A602500-250MG A602500-25G A602500-5G
77266-716EA 52 USD
77266-716 77266-718 77266-720 77266-722 77266-724
2-Methoxybenzenesulfonyl chloride 98%
2-Methoxybenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
MDL Number: MFCD01961367
CAS Number: 10130-87-7
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR