4-Chloro-6-(1H-pyrazol-1-yl)pyrimidin-5-amine 97%

Supplier: Ambeed
Danger

A601995-100MG A601995-1G A601995-250MG
77242-604EA 68 USD
77242-604 77242-606 77242-608
4-Chloro-6-(1H-pyrazol-1-yl)pyrimidin-5-amine 97%
4-Chloro-6-(1H-pyrazol-1-yl)pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₇H₆ClN₅
MDL Number: MFCD17277651
CAS Number: 1342392-11-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR