(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(3-((5-(4-fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate 97%

Supplier: Ambeed
Warning

A601674-5G A601674-25G
77254-252EA 28 USD
77254-252 77747-245
(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(3-((5-(4-fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate 97%
(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(3-((5-(4-fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate

Precaución: for Research Use Only

Formula: C₃₂H₃₃FO₉S
MDL Number: MFCD28167873
CAS Number: 866607-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR