4,7-Dimethoxy-1H-indole 97%

Supplier: Ambeed
Warning

A601630-100MG A601630-250MG
77292-788EA 40 USD
77292-788 77292-790
4,7-Dimethoxy-1H-indole 97%
4,7-Dimethoxy-1H-indole

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD00178512
CAS Number: 23876-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR