3-Cyclopropyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole 98%

Supplier: Ambeed
Warning

A601525-100MG A601525-1G A601525-250MG
77242-598EA 36.05 USD
77242-598 77242-600 77242-602
3-Cyclopropyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole 98%
3-Cyclopropyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₆H₂₁BN₂O₂
MW: 284.16 g/mol
MDL Number: MFCD27936578
CAS Number: 1650548-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR